Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LY 272015 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity 5-HT2B receptor antagonist that displays selectivity over 5-HT2A and 5-HT2C receptors (Ki values are 0.75, 21.63 and 28.7 nM for 5-HT2B, 5-HT2C and 5-HT2A receptors respectively). Orally active.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 372.89 |
Percent Purity | >99% |
Chemical Name or Material | LY 272015 hydrochloride |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction