Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY 344864 hydrochloride, Tocris Bioscience™
GSA_VA

Catalog No. p-4784124 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT1F agonist

Potent, selective 5-HT1F receptor agonist (EC50 = 3 nM). Displays > 80-fold selectivity over other 5-HT receptors (Ki values are 0.006, 0.53, 0.55, 0.56, 1.42, 1.70, 3.50, 3.94 and 4.85 μM for 5-HT1F, 5-HT1A, 5-HT1B, 5-HT1D, 5-HT1E, 5-HT2B, 5-HT2C, 5-HT2A and 5-HT7 receptors respectively). Inhibits neurogenic dural inflammation in vivo following i.v. and oral administration.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1217756-94-9
Molecular Formula C21H23ClFN3O
Molecular Weight (g/mol) 387.883
InChI Key OKUHLSYESBLBCP-PKLMIRHRSA-N
Synonym r-n-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluorobenzamide hydrochloride, ly-344864 hydrochloride, ly 344864 hydrochloride, n-6r-6-dimethylamino-6,7,8,9-tetrahydro-5h-carbazol-3-yl-4-fluorobenzamide hydrochloride, n-3r-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluoro-benzamide hcl, n-3r-3-dimethylamino-2,3,4,9-tetrahydro-1h-carbazol-6-yl-4-fluorobenzamidehydrochloride
PubChem CID 56972176
IUPAC Name N-[(6R)-6-(dimethylamino)-6,7,8,9-tetrahydro-5H-carbazol-3-yl]-4-fluorobenzamide;hydrochloride
SMILES CN(C)C1CCC2=C(C1)C3=C(N2)C=CC(=C3)NC(=O)C4=CC=C(C=C4)F.Cl

Specifications

Quantity 10 mg
Formula Weight 387.88
Percent Purity >99%
Chemical Name or Material LY 344864 hydrochloride
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.