Learn More
Ly49C Monoclonal Antibody (4LO3311), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP
Mouse Monoclonal Antibody
Supplier:  Leinco Technologies Inc L244100MG
Encompass_Preferred
Catalog No. 50-254-4137
Explore all special offers
Add to Cart
Description
Description
Specificity: 4LO3311 activity is directed against Ly49C (Killer cell lectin-like receptor 3, KLRA3; NK2.1). 4LO3311 binds an epitope located in a 32-amino acid segment of the stalk region immediately adjacent to the carbohydrate recognition domain. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -70C. Avoid Repeated Freeze Thaw Cycles.
Ly49C Monoclonal antibody specifically detects Ly49C in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays
Specifications
Specifications
Ly49C
Monoclonal
≥5.0 mg/mL
PBS with no preservative; pH 7.2-7.4
Q64329
Mouse
Protein A/G
RUO
16634
4°C short term, -80°C long term
Liquid
Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays
4LO3311
Functional Grade
Klra3
5E6; Killer cell lectin-like receptor 3; killer cell lectin-like receptor, subfamily A, member 3; Klra3; Ly49C; Ly-49c; Ly49I; Ly49I receptor; LY49I1; lymphocyte antigen 49c; natural killer cell-associated antigen 2; Nk2; Nk-2; Nk2.1; NK-2.1; T-cell surface glycoprotein Ly-49C
Immunizing 129/SvJ mice with C57BL/6 NK-enriched spleen cells
100 mg
Primary
Mouse
Antibody
IgG3 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.