Learn More
BUCHI Lyovapor™ L-300 Continuous Pro Modular Freeze Dryer
The Continuous L-300 Lyovapor™ Pro Modular is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents.
Supplier: BUCHI 11L300PH2428112004
Description
- Infinite-Technology™, allowing for continuous freeze drying due to dual condensers
- −105°C lowest condenser temperature
- Automated cleaning
- Sublimiation of both water and organic-based solvents
- Advanced software for creating and managing methods, data record and reporting
Certifications
CE Certified
Specifications
-105°C to +25°C | |
Freeze Drying | |
700 mm | |
1124.3 lb. |
Stainless Steel | |
Heatable Shelves 4, Manifold Top 12, IVC + DC Capacitive and Pirani Vacuum Sensor, Temperature Sensor, Software, Edwards nXDS10iC Vacuum Pump | |
208/230 V | |
511.1 kg |