Learn More
BUCHI Lyovapor™ L-300 Continuous Pro Modular Freeze Dryer
SDP
The Continuous L-300 Lyovapor™ Pro Modular is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents.
Supplier:  BUCHI 11L300PH2615102004
 View more versions of this product
Catalog No. 05-405-444
Description
Description
  • Infinite-Technology™, allowing for continuous freeze drying due to dual condensers
  • −105°C lowest condenser temperature
  • Automated cleaning
  • Sublimiation of both water and organic-based solvents
  • Advanced software for creating and managing methods, data record and reporting
Certifications
CE Certified
Specifications
Specifications
-105°C to +25°C
Freeze Drying
700 mm
1102.4 lb.
Stainless Steel
Heatable Shelves 6, Internal Vacuum Control (IVC), Temperature Sensor, Edwards nXDS10iC Vacuum Pump
208/230 V
501.1 kg
Product Suggestions
Product Suggestions