Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mechanix Wear™ M-Pact™ 2 Gloves - Blue
Shop All Mechanix Wear Products
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
Description
- Thermoplastic Rubber (TPR) finger guards and metacarpal protection prevent impact injuries
- Accordion knuckle guard with high density EVA foam padding
- Armortex™ fingertips provide durable gripping power
- D3O™ palm padding absorbs and dissipates high-impact energy in the palm
- TrekDry™ material helps keep hands cool and comfortable
- Extended Neoprene cuff provides added protection
- Hook and Loop closure provides a secure fit
- Machine washable
Specifications
Specifications
Cuff Material | Neoprene |
Color | Blue |
Hand | Specific |
Material | Thermoplastic Rubber |
Product Type | Mechanic's Gloves |
Size | Small |
For Use With (Application) | Construction, Heavy Equipment Operation, Maintenance & Repair, Manufacturing, Search & Rescue, and DIY Home Improvement |
Glove Cuff Style | Hook and Loop |
Series | M-Pact™ 2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction