Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ M-Pact™ 2 Gloves - Red

Catalog No. p-7214844 Shop All Mechanix Wear Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

The M-Pact™ 2 features an accordion knuckle with EVA foam padding and Thermoplastic Rubber (TPR) finger guards to protect against blunt force impact. D3O™ technology in the palms reduces impact and vibration from hand tools.

  • Thermoplastic Rubber (TPR) finger guards and metacarpal protection prevent impact injuries
  • Accordion knuckle guard with high density EVA foam padding
  • Armortex™ fingertips provide durable gripping power
  • D3O™ palm padding absorbs and dissipates high-impact energy in the palm
  • TrekDry™ material helps keep hands cool and comfortable
  • Extended Neoprene cuff provides added protection
  • Hook and Loop closure provides a secure fit
  • Machine washable

Specifications

Cuff Material Neoprene
Color Red
Hand Specific
Material Thermoplastic Rubber
Product Type Mechanic's Gloves
Size Small
For Use With (Application) Construction, Heavy Equipment Operation, Maintenance & Repair, Manufacturing, Search & Rescue, and DIY Home Improvement
Glove Cuff Style Hook and Loop
Series M-Pact™ 2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.