Learn More
Electron Microscopy Sciences MAC Rare Earth Set, 14/Set
Supplier: Electron Microscopy Sciences 8008614
This set consists of 14 rare earth compounds dispersed onto holey carbon films (3.05 mm, 400# copper grid). LaF3, CeAl2, PrF3, NdF3, SmF3, EuF3, GdF3, TbF3, DyF3, HoF3, ErF3, TmF3, YbF3, and LuF3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.