Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ MacConkey Agar
Shop All Aldon Products

Click to view available options
Quantity:
125 mL
200 mL
Description
Specifications
Specifications
Certifications/Compliance | Sterile |
Concentration | 1 |
Content And Storage | Room Temperature |
Format | Bottle |
Manufacturing Origin | United States |
Media Type | MacConkey Agar |
Product Type | Prepared Microbiology Media |
For Use With (Application) | Microbes, Educational |
Packaging Type | Glass |
Quantity | 125 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction