Learn More
Bioworld Maclura pomifera (Osage orange) Lectin (MPL/MPA) - FITC
Supplier:  Bioworld 217610391
Encompass_Preferred
Technical Specifications:Ligand Source:Maclura pomiferaMolecular Weight: 44 kDaCarbohydrate Specificity:GalNAc, Galβ1,3GalNAcInhibiting/Eluting Sugar: GalactoseCarbohydrate for Elution: 0.2M GalactoseBlood Group Specificity: A, B, O (-SA)Conjugate: FITCForm: LiquidIsoelectric (pI): 4.8-5.3Storage: -20°CApplication note: N/A
Catalog No. 50-199-154
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.