Learn More
Bioworld Maclura pomifera (Osage orange) Lectin (MPL/MPA) - Rhodamine (TRITC)
Supplier: Bioworld 217610401
Technical Specifications:Ligand Source:Maclura pomiferaMolecular Weight:44 kDaCarbohydrate Specificity:GalNAc, Galβ1,3GalNAcInhibiting/Eluting Sugar: GalactoseCarbohydrate for Elution:0.2M GalactoseBlood Group Specificity:A, B, O (-SA)Conjugate: RhodamineForm:LiquidIsoelectric (pI):4.8-5.3Storage: -20°CApplication note: N/A
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.