Learn More
Cytiva (Formerly Pall Lab) Macrosep Advance Centrifugal Devices with Omega Membrane - 10K, blue (6/pkg)
Supplier:  Cytiva MAP010C36
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Quickly Concentrates Up To 20 mL of Biological Sample-Rapidly concentrates 20 mL sample volumes to 0.5 mL-Achieve 50X concentration and > 90% recovery in just minutes-Low protein-binding Omega™ membrane and polypropylene housing minimize losses due to non-specific binding-Versatile Omega membrane is available in a variety of MWCOs-Built-in deadstop prevents spinning to dryness-Color-coded and laser etched for easy identification This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9533
missing translation for 'QuantityLeft'