Learn More
Electron Microscopy Sciences Magic Wand with Flat Fine And Flat Sharp, 5"
Supplier:  Electron Microscopy Sciences 6332303
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Magic Wands are made from PEEK, a high performance plastic, with excellent chemical resistantance and withstands high temperatures (up to 300 degree C). Magic wand is ideal for use for electronic high-tech areas to laboratories, from preparation of samples to applying adhesives, using it as a stirring rod, or evendosing samples etc.
OAL: 5"
Catalog No. 50-322-29
missing translation for 'QuantityLeft'