Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium powder, -20+100 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog No. AA00869A1
Click to view available options
Quantity:

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

7439-95-4

Molecular Formula

Mg

Molecular Weight (g/mol)

24.30

MDL Number

MFCD00085308

InChI Key

JLVVSXFLKOJNIY-UHFFFAOYSA-N

Synonym

magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs

PubChem CID

5462224

ChEBI

CHEBI:25107

IUPAC Name

magnesium

SMILES

[Mg]

Specifications

Assay Percent Range

99.8% (metals basis)

Linear Formula

Mg

Quantity

1 kg

Mesh Size

-20+100 Mesh

Chemical Name or Material

Magnesium powder

Hazard Category

H250-H260-H500

Precautionary Statement

P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c

DOTInformation

Hazard Class: 4.3; Packaging Group: II

EINECSNumber

231-104-6

RTECSNumber

OM2100000

TSCA

Yes

Recommended Storage

Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.