Learn More
Thermo Scientific MAIN CHAMBER SEAL
Supplier:  Thermo Scientific 090600600
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chamber Seal (O-Ring), Large, for Savant Speed Vac works with Savant SpeedVac Series Concentrator
Catalog No. NC9582922
missing translation for 'QuantityLeft'