Learn More
Restek™ Male Pipe to Valco™ Internal Adapter Connector

Makes a minimum volume connection from a female pipe fitting on a pressure gauge or regulator to a Valco zero-dead-volume fitting.
$28.81 - $28.86
Catalog Number | Mfr. No. | Port Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Port Type | Price | Quantity & Availability | |||||
06-710-527
![]() |
Restek™
20158 |
1/8 in. NPT male to 1/16 in. ZDV |
Each for $28.86
|
|
|||||
06-710-528
![]() |
Restek™
20159 |
1/4 in. NPT male to 1/16 in. ZDV |
Each for $28.81
|
|
|||||
Description
- 300 series stainless steel
- Includes stainless steel ferrules
- Union Bore: 1.0mm
Specifications
Connector | |
1 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.