Learn More
Gold Biotechnology Inc Mammalian Cell Lysis Buffer 500 mL
Supplier: Gold Biotechnology Inc GB180500
Mammalian Cell Lysis Buffer has been developed for extraction of total soluble proteins from mammalian cultured cells. The Mammalian Cell Lysis Buffer is based on organic buffering agents, which utilizes a mild non-ionic detergent and a proprietary combination of various salts and agents to enhance extraction and stability of proteins. Depending on the application, additional agents such as chelating agents, reducing agents and protease inhibitors may be added into Mammalian Cell Lysis Buffer. Mammalian Cell Lysis Buffer reagent has been tested for use with a wide variety of mammalian cells. Mammalian Cell Lysis Buffer can be used for both suspension as well as adherent cells. The proprietary combination of this reagent provides a simple and versatile method for the extraction of proteins from mammalian cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.