Learn More
Bioassay Systems EnzyChrom™ D-Mannitol Assay Kit. For quantitative determination of D-mannitol in biological, food, beverage and agriculture samples.
SDP
Supplier:  Bioassay Systems EMNT100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For quantitative determination of D-mannitol in biological, food, beverage and agriculture samples. Key Features: Fast and sensitive. Use of 20 µL sample. Linear detection range 0.007 to 3 mM mannitol in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Biological, food, beverage and agricultural products. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 7 µM.
Catalog No. 50-107-8347
missing translation for 'QuantityLeft'