Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Marimastat, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
Description
Broad spectrum inhibitor of MMPs (IC50 values are 3, 5, 6, 9 and 13 nM for MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 respectively). Inhibits peritoneal dissemination of human gastric cancer cells in vivo through inhibition of tumor angiogenesis.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 331.41 |
Percent Purity | >99% |
Chemical Name or Material | Marimastat |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction