Learn More
Thermo Scientific™ MAS™ Omni·Infectious™ Quality Controls
Assess the performance of Serological assays for viral and bacterial pathogens using the Thermo Scientific MAS Omni Infectious Quality Controls.
Supplier: Thermo Scientific™ 10027481
Description
Assess the performance of serological assays for viral blood stream infections using the Thermo Scientific MAS Omni Infectious Quality Controls. These third-party control panels include individual positive controls for anti-HBs, HIV-p24 and Syphilis as well as consolidated HIV, HCV, HBV and HTLV for efficiency.
- 30 days open vial stability for Syphilis; anti-HBs and HIV p24 at 2 to 8°C
- Ready to use straight from the refrigerator
- Human serum matrix
- BSI Positive and Infectious Negative
- BSI Positive has Multiple analytes in a single vial
Specifications
2°C to 8°C | |
HIV-p24, HIV, HCV, HBV, HTLV | |
One box of control contains five tubes of HIV p24 Positive control |
MAS HIV p24 Positive Control | |
Control | |
5 x 5.0 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.