Learn More
MasterTech 100% Reagent Alcohol, StatLab™

- An effective, safe, alternative option
- No procedure changes on the user's end
- Formulated using 95% 200 proof Ethyl Alcohol and 5% Isopropyl Alcohol
Specifications
Physical Form
Aqueous Solution
Color
Clear
Quantity
1 gal./Bottle, 4 Bottles/Cs.
Grade
Reagent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.