Learn More
Verichem Laboratories Reference Kit Matrix Plus™ Chemistry 1 X 15 mL
Protein based, ready-to-use liquid. No glycols, azides or surfactants. Known concentrations verified against available NIST standard reference material. Glucose, Urea Nitrogen, Creatinine, Calcium, Phosphorus, Magnesium, Triglyceride Analytes