Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maxi Potassium channel alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | Maxi Potassium channel alpha |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Maxi Potassium channel alpha Polyclonal specifically detects Maxi Potassium channel alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Maxi Potassium channel alpha | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q12791 | |
3778 | |
This antibody was developed against a recombinant protein corresponding to amino acids: INLCDMCVILSANQNNIDDTSLQDKECILASLNIKSMQFDDSIGVLQANSQGFTPPGMDRSSPDNSPVHGMLRQPSITTGVNIPIITELVNDTNVQFLDQDDDDDPDTELYLTQP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Potassium Channels | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
bA205K10.1, BK channel, BKCA alpha, BKCA alpha subunit, BKTM, calcium-activated potassium channel subunit alpha-1, DKFZp686K1437, hSlo, k(VCA)alpha, KCNMA, Maxi K channel, MaxiK, MGC71881, potassium large conductance calcium-activated channel, subfamily M, alphamember 1, SAKCA, Slo homolog, Slo1, SLO-ALPHA, Slowpoke homolog, stretch-activated Kca channel, subfamily M subunit alpha-1 | |
KCNMA1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title