Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Maxi Potassium channel beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$173.10
Specifications
Antigen | Maxi Potassium channel beta |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 2.0 μg/mL, Flow Cytometry, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:200 - 1:2000, Knockdown Validated |
Applications | Western Blot |
Classification | Polyclonal |
Description
Maxi Potassium channel beta Polyclonal specifically detects Maxi Potassium channel beta in Human, Mouse, Rat, Porcine, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Maxi Potassium channel beta | |
Western Blot 2.0 μg/mL, Flow Cytometry, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:200 - 1:2000, Knockdown Validated | |
Polyclonal | |
Rabbit | |
Potassium Channels | |
PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
BK channel beta subunit, BK channel subunit beta-1, BKbeta, BKbeta1, Calcium-activated potassium channel subunit beta, calcium-activated potassium channel subunit beta-1, Calcium-activated potassium channel, subfamily M subunit beta-1, Charybdotoxin receptor subunit beta-1, Hbeta1, hslo-beta, K(VCA)beta, k(VCA)beta-1, large conductance Ca2+-activated K+ channel beta 1 subunit, Maxi K channel beta subunit, Maxi K channel subunit beta-1, potassium large conductance calcium-activated channel, subfamily M, beta member1, Slo-beta, Slo-beta-1 | |
KCNMB1 | |
IgG | |
Affinity Purified |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Q16558 | |
3779 | |
Synthetic Peptide: L(90) Y H T E D T R D Q N Q Q C(103) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title