Learn More
Apexbio Technology LLC mCAP, 10ul (100 mM). Cas: MFCD: NA. m7G(5')ppp(5')G Cap Analog, used to forming Cap 0 structure
SDP
Supplier:  Apexbio Technology LLC B8174.01
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
mRNA of eucaryon and virus has a monomethylated cap structure, m7G(5')ppp(5')G or called mCAP, at the 5-initiating terminal. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5477