Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kodiak™ McKinney CT CP ESR WP 6 in. Brown Boot

Composite safety-toe construction combines with the composite safety plate to provide essential protection on the job-site. With seam-sealed waterproof construction and superior traction on everything from slick concrete to loose gravel, they’ll keep you dry and safe in every season.

Supplier:  Kodiak™ K4TFBN 110 W

 View more versions of this product

Catalog No. 17-100-171


Add to Cart

Description

Description

  • Classic Kodiak™ 6 in. boot design with bellows leather tongue and padded collar with integrated pull tab
  • Full grain waterproof leather with waterproof membrane construction
  • Kodiak™ SOS Smell Out Science™ odor control technology
  • Abrasion resistant lining
  • Comfortzone™ footbed with Ortholite™ comfort foam
  • Slip resisting, non-marking rubber outsole
Specifications

Specifications

Brown
11W
Yes
Boot
Leather
Men's
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.