Learn More
MCR Safety Blackjack™ Safety Glasses
Deal your workers the Blackjack™ from MCR Safety. With these sleek, style-conscious safety glasses, everybody wins. Blackjack features a frameless lens design with metal temples and non-slip temple sleeves.
Supplier: MCR Safety S21150
Description
- Rimless design for unobstructed vision
- Fully adjustable metal alloy hybrid temples
- Non-slip, soft temple material
- Universal nose bridge
- Welding styles available
Specifications
IR Filter Shade 5.0 | |
12/Pk. | |
Chrome |
144/Cs. | |
Polycarbonate |