Learn More
Electron Microscopy Sciences SEM Medium Resolution Au on Carbon Test Specimen, Hitachi Stub
Supplier:  Electron Microscopy Sciences 7951604
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Larger gold particles in this test with wider gaps, allow for structured microscope setup. Magnification range is 800-10,000x.
Type: Hitachi Stub
Catalog No. 50-193-1212
missing translation for 'QuantityLeft'