Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mefenamic Acid, BP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18604272 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
Packaging:

M1117, 61-68-7, C15H15NO2

Spectrum™ Chemical Mefenamic Acid, BP is used for the short-term treatment of mild to moderate pain from various conditions. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS

61-68-7

Molecular Formula

C15H15NO2

Molecular Weight (g/mol)

241.29

InChI Key

HYYBABOKPJLUIN-UHFFFAOYSA-N

IUPAC Name

2-[(2,3-dimethylphenyl)amino]benzoic acid

SMILES

CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C

Specifications

CAS Max %

100%

Loss on Drying

0.5%

Packaging

Poly Pail

Quantity

5 kg

Percent Purity

99 to 101%

Grade

BP

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.