Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mefenamic Acid, BP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. p-7157710 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Pail

M1117, 61-68-7, C15H15NO2

Spectrum™ Chemical Mefenamic Acid, BP is used for the short-term treatment of mild to moderate pain from various conditions. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 61-68-7
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
InChI Key HYYBABOKPJLUIN-UHFFFAOYSA-N
IUPAC Name 2-[(2,3-dimethylphenyl)amino]benzoic acid
SMILES CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Poly Pail
Quantity 1 kg
Percent Purity 99 to 101%
Grade BP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.