Learn More
TARGETMOL CHEMICALS INC MEFENIDIL 5MG
Supplier:  TARGETMOL CHEMICALS INC T33272
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Mefenidil (McN-2378) a selective cerebral vasodilator was shown to increase CBF in healthy brains without stimulating O2 uptake in a dog anesthesia model. purity: 98%
Catalog No. 50-258-0315
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.