Learn More
Cytiva GLA-5000 PVC Membrane Disc Filters - 5 m, 47 mm (100/pkg)
Supplier:  Cytiva 66468
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Inherently Low Ash Membrane Ideally Suited for Multiple NIOSH Analytical Methods-Assures gravimetric stability with low moisture pick-up and low tare weight.-Low ash. Provides interference-free silica determinations.-5 μm pore size meets NIOSH and OSHA requirements.-25 and 37 mm sizes are ideal for use in Pall Air Monitoring Cassettes. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3326