Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Memory Foam Antifatigue Insoles

Impacto™ Memory Foam Antifatigue Insoles are designed to provide comfort, impact absorption and support to help reduce fatigue when standing on hard surfaces for long periods.

Supplier:  Impacto™ MEM1213

 View more versions of this product

Catalog No. 19-400-128


missing translation for 'QuantityLeft'
Add to Cart

Description

Description

Impacto™ Memory foam Anti-Fatigue Insoles

  • Comfort to the natural curves of foot for a customized fit
  • Offer impact protection when walking or running
  • Molded style for arch and heel support
  • Ideal for safety footwear, boots and running shoes
  • Soft fabric cover to eliminate moisture
  • Antifungal and antibacterial materials
  • Can be trimmed for a perfect fit
  • PVC-free
Specifications

Specifications

Orange, Black
30.48 cm
Men: 12 to 13, Women: 14
Unisex
12 in.
Memory Foam
Molded
Memory foam insoles
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.