Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Menthol
SCI_ED

Catalog No. p-4443170 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Solid

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
InChI Key NOOLISFMXDJSKH-IVZWLZJFSA-N
Synonym menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
PubChem CID 6566020
IUPAC Name (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Color White
Density 0.890g/cmsuperscript_3;
Assay Percent Range 99% w/v
Packaging Solid
Linear Formula C10H20O
Quantity 25 g
Formula Weight 156.26
Chemical Name or Material Menthol
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.