Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury AA Standard, 1000 ppm Hg in 3% HNO3, Ricca Chemical
SDP

Catalog No. p-3251921 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 21908-53-2, 7697-37-2, 7732-18-5
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Packaging Natural LDPE Bottle
Quantity 100 mL
CAS Min % 93.0, 4.63, 0.1
CAS Max % 97.0, 4.81, 0.1
Color Colorless
Concentration or Composition Notes 1,000ppm Hg
Equivalence 1 mL = 1 mg Hg
Physical Form Liquid
Chemical Name or Material Mercury AA Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.