Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury, ACS, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. p-3257048
Click to view available options
Quantity:
0.5 lb.
1 lb.
5 lb.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 7439-97-6
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
Synonym hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]

Specifications

Color Silver
Appearance Silver-colored liquid
Density 13.534g/cm3 at 20°C
Assay Percent Range 99.999% (Metals basis)
Quantity 0.5 lb.
Formula Weight 200.59
Grade ACS
Odor Odorless
Vapor Pressure 0.00163 hPa
Form Liquid
Chemical Name or Material Mercury
Show More Show Less
Hazard Category H330-H360D-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-106-7
RTECSNumber OV4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.