Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) bromide, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC448121000
Click to view available options
Quantity:
Packaging:

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

7789-47-1

Molecular Formula

Br2Hg

Molecular Weight (g/mol)

360.4

InChI Key

NGYIMTKLQULBOO-UHFFFAOYSA-L

Synonym

mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide

PubChem CID

24612

ChEBI

CHEBI:49639

IUPAC Name

dibromomercury

SMILES

Br[Hg]Br

Specifications

Boiling Point

322.0°C

Melting Point

236.0°C

Quantity

100 g

Linear Formula

HgBr2

Merck Index

15,5942

Solubility Information

Solubility in water: 0.6g/100mL (25 c). Other solubilities: freely soluble in hcl and hbr, soluble in alcohol and ether, slightly soluble in chloroform

Formula Weight

360.4

Grade

ACS Reagent

Packaging

Glass bottle

Chemical Name or Material

Mercury(II) bromide

Hazard Category

GHS Signal Word: Danger

EINECSNumber

232-169-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.