Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) bromide, ACS, Thermo Scientific Chemicals

Catalog No. p-4407638
Click to view available options
Quantity:
25 g
100 g

Mercury(II) bromide is used as a reagent in the Koenigs-Knorr reaction, which forms glycoside linkages on carbohydrates. It is also used to test for the presence of arsenic, as recommended by the Pharmacopoeia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-47-1
Molecular Formula Br2Hg
Molecular Weight (g/mol) 360.4
MDL Number MFCD00011040
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
PubChem CID 24612
ChEBI CHEBI:49639
IUPAC Name dibromomercury
SMILES Br[Hg]Br

Specifications

Boiling Point 322°C to 325°C (sublimation)
Melting Point 236°C
Color White
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in hot alcohol,methanol,HCl,HBr
Formula Weight 360.4
Grade ACS
Odor Odorless
Sensitivity Light sensitive
Vapor Pressure 1mm Hg at 137°C
Density 6.05 g/cm3 at 20°C
Chemical Name or Material Mercury(II) bromide, For ACS analysis
Show More Show Less
Hazard Category H300+H310+H330-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 232-169-3
RTECSNumber OV7415000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.