Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals

Catalog No. AAA1633030
Click to view available options
Quantity:

It is used in medicine as a catalyst and in extracting gold and silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine as a catalyst and in extracting gold and silver.

Solubility
It is soluble in hot H2SO4, NaCl solution. insoluble in alcohol, acetone, ammonia

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Light sensitive. Moisture sensitive. Air sensitive. Keep away from Strong oxidizing agents.

Chemical Identifiers

CAS

7783-35-9

Molecular Formula

HgO4S

Molecular Weight (g/mol)

296.65

MDL Number

MFCD00011047

InChI Key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Synonym

mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate

PubChem CID

24544

SMILES

[Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point

(decomposition)

Quantity

250 g

Linear Formula

HgSO4

UN Number

UN1645

Merck Index

14,5887

Solubility Information

It is soluble in hot H2SO4,NaCl solution. insoluble in alcohol,acetone,ammonia

IUPAC Name

mercury(2+) sulfate

Formula Weight

296.65

Percent Purity

≥98%

Sensitivity

Moisture sensitive

Density

6.47

Chemical Name or Material

Mercury(II) sulfate

Hazard Category

H300+H310+H330-H373

Hazard Statement

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

Precautionary Statement

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c

DOTInformation

Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY SULFATES

EINECSNumber

231-992-5

RTECSNumber

OX0500000

TSCA

Yes

Recommended Storage

Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.