Learn More
Mercury(II) trifluoroacetate, 98+%, Thermo Scientific Chemicals
Description
Mercury(II) trifluoroacetate promotes selective cyclization of methylthio-substituted divinyl ketones to 5-fluorocyclopentenone derivatives. It is a raw material of fine chemicals and pharmaceutical intermediates. So it can be used in the pharmaceutical synthesis. Both linear and cross-linked copolymers of N-vinylcarbazole and methylacrylate can be prepared and mercuriated by using mercury trifluoroacetate in THF or dichloromethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 167°C to 169°C |
Color | White |
Linear Formula | Hg(OOCCF3)2 |
Quantity | 5 g |
UN Number | UN2025 |
Beilstein | 3633860 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 426.62 |
Percent Purity | ≥98% |
Show More |
Safety and Handling
Hazard Category | H300+H310+H330-H373 |
Hazard Statement | GHS H Statement H300-H310-H330-H373 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S. |
EINECSNumber | 236-250-4 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.