Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
meso-Butane-1,2,3,4-tetracarboxylic Acid 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
500 g
Specifications
Specifications
Color | White-Yellow |
Quantity | 25 g |
Formula Weight | 234.16 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | meso-Butane-1,2,3,4-tetracarboxylic Acid |
Safety and Handling
EINECSNumber | (2)-0897&(2)-1485 |
RTECSNumber | EK6100000 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction