Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meta-Phosphoric acid, puriss p.a., ≥33.5% (T), Honeywell™ Fluka™

Catalog No. p-7120569 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Color Colorless
Melting Point 600°C
Packaging Poly bottle
Quantity 100 g
Physical Form Solid
Linear Formula (HPO3)n
UN Number UN3260
Percent Purity ≥33.5% (T)
Chemical Name or Material meta-Phosphoric acid
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8 / PGIII
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.