Learn More
Cole-Parmer Simpson METER0-1DCMA 4.5 DSP
Supplier: Cole-Parmer 6848967
FEATURES
- 3-1/2 (F35) or 4-1/2 (F45) digit, 0.5" High Red LED Display
- Front Panel Pops Off For Quick Changes in Decimal Point of Display
- Scaling
- 120 VAC Power Supply
- Application Flexibility - inputs available for DC, AC, or ACTRMS voltage or current, temperature, or frequency
- 9-32 VDC Power Available (option)
- Panel Cutout: 1.77" x 3.62" Depth: 3.12"