Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrylic Acid, Spectrum™ Chemical
SDP

Catalog No. p-7226022 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL

Synonym: 2-Methylacrylic Acid, Polymethacrylic Acid, 2-Methylpropenoic Acid, Alpha-Methylacrylic Acid

Methacrylic Acid is used as a building block to make a wide range of polymer-based products like binders in paints and coatings, water-soluble polymers and interlevant in glass. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 79-41-4

Specifications

Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.