Learn More
Methaqualone-D7 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
Shop All MilliporeSigma ProductsMethaqualone is a sedative-hypnotic and a central nervous system depressant sold under the trade name Quaalude for treatment of insomnia. Methaqualone also has a history of abuse, known by street names such as Disco biscuits, Lemmons, and 714s. This internal standard is suitable for quantitation of methaqualone levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology or forensic analysis.
Specifications
Chemical Name or Material
Methaqualone-D7
Concentration
100 μg/mL in Methanol
CAS
136765-41-8
Packaging
Ampule of 1 mL
Solution Type
Standard Solution
Type
Certified Reference Material
For Use With (Equipment)
Gas Chromatography (GC), Liquid Chromatography (LC)
Molecular Formula
C16H7D7N2O
Molecular Weight (g/mol)
257.34
Grade
Certified
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.