Learn More
Medchemexpress LLC HY-B0553 500mg Medchemexpress, Methazolamide CAS:554-57-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0553500MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-B0553 500mg Methazolamide CAS:554-57-4 Methazolamide (L584601) is a sulfonamide derivative used as a carbonic anhydrase inhibitor with a Ki of 14 nM for human carbonic anhydrase II. Methazolamide, an intraocular pressure-lowering agent, reduces intraocular pressure elevations associated with glaucoma and other ocular disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2597
missing translation for 'QuantityLeft'