Learn More
Nitrogen-Phosphorous Pesticides Mix D, SPEX CertiPrep™
Shop All Spex CertiPrep Products

Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Beam | 41814-78-2 | 200μg/mL |
Fenarimol | 60168-88-9 | 200μg/mL |
Norflurazon | 27314-13-2 | 200μg/mL |
Phenamiphos | 22224-92-6 | 200μg/mL |
Sonar/Flouridone | 59756-60-4 | 200μg/mL |
Tebuthiuron | 34014-18-1 | 200μg/mL |
Tetrachlorvinphos | 961-11-5 | 200μg/mL |
Triadimefon | 43121-43-3 | 200μg/mL |
Velpar | 51235-04-2 | 200μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Chemical Name or Material
Nitrogen-Phosphorus Pesticides Mix D
Packaging
2mL amber prescored glass ampule
Quantity
1 mL
Concentration or Composition Notes
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment)
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Methods
For US EPA Method 525.2
Analyte or Component Names
Beam; Fenarimol; Norflurazon; Phenamiphos; Sonar/Flouridone; Tebuthiuron; Tetrachlorvinphos; Triadimefon; Velpar
Solvent or Matrix
Acetone
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.