Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Method 525.2 Nitrogen-Phosphorous Pesticides Mix E, SPEX CertiPrep™
SDP

Catalog No. 5252E Shop All Spex CertiPrep Products
Click to view available options
Quantity:
Packaging:

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Carboxin5234-68-4200μg/mL
Disulfoton298-04-4200μg/mL
Tributylphosphoro-trithioite150-50-5200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material

Nitrogen-Phosphorus Pesticides Mix E

Packaging

2mL amber prescored glass ampule

Quantity

1 mL

Concentration or Composition Notes

Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.

For Use With (Equipment)

GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation

Methods

For US EPA Method 524

Analyte or Component Names

Carboxin; Disulfoton; Tributylphosphoro-trithioite

Solvent or Matrix

Acetone

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.