Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxy-X04, Tocris Bioscience™

Catalog No. p-4817134 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorescent amyloid β detector; brain penetrant

Methoxy-X04 is a fluorescent amyloid β (Aβ) probe for the detection and quantification of plaques, tangles and cerebrovascular amyloid. Methoxy-X04 Displays high in vitro binding affinity (Ki = 26.8 nM); binds selectively to fibrillar β-sheet deposits. Methoxy-X04 is a derivative of Congo Red (Cat. No. 5167). Methoxy-X04 can be used for in vivo Aβ plaque labeling. Methoxy-X04 co-localizes with CD68+ phagosomes in plaque-associated Iba1+ microglia; and it labels molecularly distinct plaque-associated microglia populations. Brain penetrant.

Chemical Identifiers

CAS 863918-78-9
Molecular Formula C23H20O3
Molecular Weight (g/mol) 344.41
InChI Key FGYNZFHVGOFCMD-KHVHPYDTSA-N
Synonym methoxy-x04, methoxy-04, 4,4'-2-methoxy-1,4-phenylenebisvinylene bisphenol, 4,4'-2-methoxy-1,4-phenylene di-1e-2,1-ethenediyl bisphenol
PubChem CID 16049314
IUPAC Name 4-[(E)-2-[4-[(E)-2-(4-hydroxyphenyl)ethenyl]-3-methoxyphenyl]ethenyl]phenol
SMILES COC1=C(C=CC(=C1)C=CC2=CC=C(C=C2)O)C=CC3=CC=C(C=C3)O

Specifications

Quantity 10 mg
Formula Weight 344.4
Percent Purity >98%
Chemical Name or Material Methoxy-X04
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.