Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

methyl 2-[1-methyl-3-(trifluoromethyl)-1h-pyrazol-5-yl]benzoate, 90%, Thermo Scientific™

Catalog No. CC62122DE
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

898289-62-8

Molecular Formula

C13H11F3N2O2

Molecular Weight (g/mol)

284.238

MDL Number

MFCD09879929

InChI Key

VBXXEOGWKRJBBH-UHFFFAOYSA-N

Synonym

methyl 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzoate, methyl 2-2-methyl-5-trifluoromethyl pyrazol-3-yl benzoate, 2-2-methyl-5-trifluoromethyl-3-pyrazolyl benzoic acid methyl ester, 5-2-methoxycarbonyl phenyl-1-methyl-3-trifluoromethyl-1h-pyrazole, benzoic acid,2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl-, methyl ester

PubChem CID

24229663

IUPAC Name

methyl 2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]benzoate

SMILES

CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2C(=O)OC

Specifications

CAS Min %

89.5

CAS Max %

100.0

Assay Percent Range

90%

Packaging

Amber Glass Bottle

Quantity

5 g

Formula Weight

284.24

Percent Purity

90%

Chemical Name or Material

Methyl 2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoate

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

Precautionary Statement

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.